4-Chloro-3-fluorobenzaldehyde ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C1506-1G C1506-25G C1506-5G
TCIAC1506-1GEA 26.4 EUR
TCIAC1506-1G TCIAC1506-25G TCIAC1506-5G
4-Chloro-3-fluorobenzaldehyde ≥97.0% (by GC)
4-Chloro-3-fluorobenzaldehyde
Formule: C₇H₄ClFO
MW: 158.56 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00143288
CAS nummer: 5527-95-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ecc1102a007e11c696612722143de63ce081fe8&vid=a769f30533d7612c27ef32d2c32365de14256ec4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 46.0 to 50.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR