2-Chloro-6-fluorobenzonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C1002-25G C1002-5G
TCIAC1002-25GEA 52.8 EUR
TCIAC1002-25G TCIAC1002-5G
2-Chloro-6-fluorobenzonitrile ≥98.0% (by GC)
2-Chloro-6-fluorobenzonitrile
Formule: C₇H₃ClFN
Kookpunt: 226 °C (1013 hPa)
Smeltpunt: 58…59 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001780
CAS nummer: 668-45-1
EINECS: 211-571-2
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5a20e30a9198a9b15e4fd0994cdb790c5240801&vid=9da1ddc6e6cc696e20586b7b49c97c295b8489f9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C