2-Chloro-4-fluorobenzonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
C2129-25G C2129-5G
TCIAC2129-25GEA 121 EUR
TCIAC2129-25G TCIAC2129-5G
2-Chloro-4-fluorobenzonitrile ≥98.0% (by GC)
2-Chloro-4-fluorobenzonitrile
Formule: C₇H₃ClFN
Kookpunt: 237 °C (1013 hPa)
Smeltpunt: 63…65 °C
MDL Nummer: MFCD00042523
CAS nummer: 60702-69-4
EINECS: 262-384-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42afc2517ea6230bf5db7e0c5f798aa4b02e1107&vid=e3b23fb6be90aab2557bd2df5c16bfbdd5f0395b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C