3-Chloro-4-fluoroaniline ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C1024-25G
C1024-500G
TCIAC1024-25GEA
17.6
EUR
TCIAC1024-25G
TCIAC1024-500G
3-Chloro-4-fluoroaniline ≥98.0% (by GC)
3-Chloro-4-fluoroaniline
Formule:
C₆H₅ClFN MW: 145.56 g/mol Kookpunt: 230 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007767 CAS nummer: 367-21-5 EINECS: 206-682-8 UN: 2811 ADR: 6.1 III |
|
Specification Test Results
Appearance | Gray to Dark purple to Black powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 45.0 to 47.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...