2,6-Dichlooraniline ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D0323-25G D0323-500G D0323-100G
TCIAD0323-25GEA 18.7 EUR
TCIAD0323-25G TCIAD0323-500G TCIAD0323-100G
2,6-Dichlooraniline ≥99.0% (by GC)
2,6-Dichlooraniline
Formule: C₆H₅Cl₂N
Kookpunt: 220 °C (1013 hPa)
Smeltpunt: 35…38 °C
Dichtheid: 1,526 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007675
CAS nummer: 608-31-1
EINECS: 210-160-5
UN: 3442
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c37b1ad4fb12801031a0f9a1acaddf0eb72c9f93&vid=caf7d2950d98a60e57162b19ea33368e020c2047&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 37.0 to 40.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR