3,5-Dichlooraniline ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D0325-25G D0325-500G D0325-100G
TCIAD0325-25GEA 50.6 EUR
TCIAD0325-25G TCIAD0325-500G TCIAD0325-100G
3,5-Dichlooraniline ≥98.0% (by GC)
3,5-Dichlooraniline
Formule: C₆H₅Cl₂N
Kookpunt: 260°C
Smeltpunt: 51 °C
Dichtheid: 1,58 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007774
CAS nummer: 626-43-7
EINECS: 210-948-9
UN: 3442
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e3ebff94a4c64e0fd999471d7d7960d27121bcc&vid=b4f0c6b860b18c175f81b3d0efaf72f79e9a64f0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 53.0 deg-C