4-Chlooraniline ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C0112-25G C0112-500G
TCIAC0112-25GEA 17.6 EUR
TCIAC0112-25G TCIAC0112-500G
4-Chlooraniline ≥99.0% (by GC)
4-Chlooraniline
Formule: C₆H₆ClN
MW: 127.57 g/mol
Kookpunt: 232
Vlampunt: 120
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007835
CAS nummer: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1 II
Merck Index: 14,02118
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64db6904619cb3b029dc20bc7c4747c26a9e0305&vid=b91f0ed330fd3dee583aeccf7034eaea716fb570&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 99.0 %
Melting point 69.0 to 73.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR