4-Chloor-2-nitroaniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C0215-25G C0215-500G
TCIAC0215-25GEA 19.8 EUR
TCIAC0215-25G TCIAC0215-500G
4-Chloor-2-nitroaniline ≥98.0% (by GC)
4-Chloor-2-nitroaniline
Formule: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Dichtheid: (anhydr.)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007836
CAS nummer: 89-63-4
EINECS: 201-925-4
UN: 2237
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=162eb6bd1ec0b2decfb7cd1c67244ac6834b4ce9&vid=ece0bf295d9a440f5d2141d1426486b2af3cde90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 118.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR