1,2-Dichloro-4-nitrobenzene ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D0388-25G D0388-500G
TCIAD0388-25GEA 17.6 EUR
TCIAD0388-25G TCIAD0388-500G
1,2-Dichloro-4-nitrobenzene ≥97.0% (by GC)
1,2-Dichloro-4-nitrobenzene
Formule: C₆H₃Cl₂NO₂
Kookpunt: 255°C
Smeltpunt: 38…41 °C
Dichtheid: 1,4558 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007207
CAS nummer: 99-54-7
EINECS: 202-764-2
UN: 1578
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=860c1045c27f4f11f2b9d437ee8202252f66cfd9&vid=74a2de155b5c5586e132f7da35a84e7dfc40f33e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 38.0 to 43.0 deg-C