3-Chloorfenol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C0242-25G C0242-500G C0242-100G
TCIAC0242-25GEA 28.6 EUR
TCIAC0242-25G TCIAC0242-500G TCIAC0242-100G
3-Chloorfenol ≥98.0% (by GC)
3-Chloorfenol
Formule: C₆H₅ClO
MW: 128.56 g/mol
Kookpunt: 214
Vlampunt: 97
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002256
CAS nummer: 108-43-0
EINECS: 203-582-6
UN: 2020
ADR: 6.1 III
Merck Index: 14,02154
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0e61e5399d8953fc6d5cf01acf8d1cc160435ea&vid=a23df139a43e6d6901e08a109b40d1bffad0fce4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %