4-Chlorothiophenol ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C0129-25G C0129-500G
TCIAC0129-25GEA 56.1 EUR
TCIAC0129-25G TCIAC0129-500G
4-Chlorothiophenol ≥98.0% (by GC, titration analysis)
4-Chlorothiophenol
Formule: C₆H₅ClS
Kookpunt: 206°C
Smeltpunt: 49…53 °C
Dichtheid: 1,224 g/cm³ (60 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004847
CAS nummer: 106-54-7
EINECS: 203-408-9
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04f1607e57d9a876c0562147af9bd40f8c5e598f&vid=30f02ff4fcf582107970594bbf688a914721d568&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %
Melting point 51.0 to 55.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR