1,2,4,5-Tetrafluorobenzene ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
T1029-25G
T1029-5G
TCIAT1029-25GEA
142
EUR
TCIAT1029-25G
TCIAT1029-5G
1,2,4,5-Tetrafluorobenzene ≥99.0% (by GC)
1,2,4,5-Tetrafluorobenzene
Formule:
C₆H₂F₄ Kookpunt: 90°C Smeltpunt: 4 °C Dichtheid: 1,42 g/cm³ (20 °C) Vlampunt: 16°C |
MDL Nummer:
MFCD00000307 CAS nummer: 327-54-8 EINECS: 206-319-3 UN: 1993 ADR: 3,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...