2-Fluorobenzaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
F0190-25ML F0190-250ML
TCIAF0190-25MLEA 31.9 EUR
TCIAF0190-25ML TCIAF0190-250ML
2-Fluorobenzaldehyde ≥98.0% (by GC)
2-Fluorobenzaldehyde
Formule: C₇H₅FO
MW: 124.11 g/mol
Kookpunt: 175°C
Smeltpunt: –45°C
Vlampunt: 55°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003302
CAS nummer: 446-52-6
EINECS: 207-171-2
UN: 1989
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b80e2b9ea627d3626efaab409c1080d77d5f3101&vid=4a77ed761a2f34f86389e938a7d0235da0ccaeaf&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %