3-Fluorobenzaldehyde ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
F0189-25ML F0189-5ML
TCIAF0189-25MLEA 44 EUR
TCIAF0189-25ML TCIAF0189-5ML
3-Fluorobenzaldehyde ≥97.0% (by GC)
3-Fluorobenzaldehyde
Formule: C₇H₅FO
MW: 124.11 g/mol
Kookpunt: 173°C
Vlampunt: 56°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003348
CAS nummer: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37f2575585c48dddaaffe119e886f0a1ce9b50dd&vid=636535ca2901172ba797bac1ee672470e055d560&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %