2,4-Difluorobenzoic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D1833-25G D1833-500G
TCIAD1833-25GEA 60.5 EUR
TCIAD1833-25G TCIAD1833-500G
2,4-Difluorobenzoic acid ≥98.0% (by titrimetric analysis)
2,4-Difluorobenzoic acid
Formule: C₇H₄F₂O₂
Kookpunt: 241 °C (1013 hPa)
Smeltpunt: 185 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011670
CAS nummer: 1583-58-0
EINECS: 216-430-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21cb89baee89b0642dd4bc1a6c56fd84d654c000&vid=5bf46d57ecd29bdb56ce95d87cffea782b1372ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light red powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 185.0 to 189.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR