1-Fluoro-2-iodobenzene ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Fluoroiodobenzene
F0253-10G F0253-25G
TCIAF0253-10GEA 19.8 EUR
TCIAF0253-10G TCIAF0253-25G
1-Fluoro-2-iodobenzene ≥99.0% (by GC)
1-Fluoro-2-iodobenzene
Formule: C₆H₄FI
MW: 222 g/mol
Kookpunt: 189°C
Smeltpunt: -41.5
Vlampunt: 71°C
MDL Nummer: MFCD00001031
CAS nummer: 348-52-7
EINECS: 206-477-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cc6d81bd15580b3acfb22d4f88f59daec1313a4&vid=49a1e16851f5d1da46b240d41323c45ec14a4dd9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 99.0 %
Appearance Colorless to Orange to Green clear liquid