2,5-Difluoroaniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D1634-25G D1634-5G
TCIAD1634-25GEA 89.1 EUR
TCIAD1634-25G TCIAD1634-5G
2,5-Difluoroaniline ≥98.0% (by GC)
2,5-Difluoroaniline
Formule: C₆H₅F₂N
Kookpunt: 174°C
Smeltpunt: 13 °C
Dichtheid: 1,285 g/cm³ (20 °C)
Vlampunt: 68°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007651
CAS nummer: 367-30-6
EINECS: 206-690-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=229c3459d6d3e8432652b6f9a3b53ff47f815083&vid=70964d1ae63abc4ebf462e9916f1134742418745&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR