1,3-Difluoro-5-nitrobenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D3426-25G D3426-5G
TCIAD3426-25GEA 349 EUR
TCIAD3426-25G TCIAD3426-5G
1,3-Difluoro-5-nitrobenzene ≥98.0% (by GC)
1,3-Difluoro-5-nitrobenzene
Formule: C₆H₃F₂NO₂
Kookpunt: 177°C
Vlampunt: 73°C
MDL Nummer: MFCD00012142
CAS nummer: 2265-94-3
EINECS: 218-867-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abd2a30fbfb78e387800b080b2b3f54f14674fb2&vid=36abb61c4f2b64c1bd77241a6448001da3ea5ad2&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %