1-Fluoro-4-nitrobenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
F0105-25G F0105-500G F0105-100G
TCIAF0105-25GEA 17.6 EUR
TCIAF0105-25G TCIAF0105-500G TCIAF0105-100G
1-Fluoro-4-nitrobenzene ≥98.0% (by GC)
1-Fluoro-4-nitrobenzene
Formule: C₆H₄FNO₂
Kookpunt: 207°C
Smeltpunt: 21 °C
Dichtheid: 1,326 g/cm³ (20 °C)
Vlampunt: 83 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007282
CAS nummer: 350-46-9
EINECS: 206-502-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b26ce6aefeffe965d08bd599fe8da09322096e30&vid=b7e1181995ec1c90c7082cbbc3f65eac3ad8f9f5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Yellow to Green powder to lump to clear liquid
Purity (GC) min. 98.0 %