2,6-Difluorophenol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D2015-25G D2015-5G
TCIAD2015-25GEA 223 EUR
TCIAD2015-25G TCIAD2015-5G
2,6-Difluorophenol ≥98.0% (by GC)
2,6-Difluorophenol
Formule: C₆H₄F₂O
Kookpunt: 60°C
Smeltpunt: 46 °C
Dichtheid: 1,27 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002158
CAS nummer: 28177-48-2
EINECS: 248-884-9
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84dea3ae9c48c3ac335c0a42d81a7ee08a78c387&vid=157db0c0d861ff66f2884750a7ef37aceae198f9&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency