2-Fluorophenol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

F0150-25G F0150-250G
TCIAF0150-25GEA 19.8 EUR
TCIAF0150-25G TCIAF0150-250G
2-Fluorophenol ≥98.0% (by GC)
2-Fluorophenol
Formule: C₆H₅FO
Kookpunt: 152°C
Smeltpunt: 16 °C
Dichtheid: 1,22 g/cm³ (20 °C)
Vlampunt: 47°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002155
CAS nummer: 367-12-4
EINECS: 206-681-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3376f687c7d90f61161fddb4c9c33041b590ee46&vid=874acab580ab0140ff2a1f447c0937af3936a7a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR