Iodobenzene ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Phenyliodide
I0050-25G I0050-500G I0050-100G
TCIAI0050-25GEA 19.8 EUR
TCIAI0050-25G TCIAI0050-500G TCIAI0050-100G
Iodobenzene ≥99.0% (by GC)
Iodobenzene
Formule: C₆H₅I
Kookpunt: 188°C
Smeltpunt: –29°C
Dichtheid: 1,838 g/cm³ (20 °C)
Vlampunt: 74°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001029
CAS nummer: 591-50-4
EINECS: 209-719-6
Merck Index: 14,05029
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97c144a3cd20c73544b4ee4c31536141d065680a&vid=cb8a7cd5aca6d2e369927b47f5b618087aab2483&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity (GC) min. 99.0 %
Appearance Colorless to Light orange to Yellow clear liquid