1-Iodo-4-nitrobenzene ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
I0064-25G I0064-100G
TCIAI0064-25GEA 62.7 EUR
TCIAI0064-25G TCIAI0064-100G
1-Iodo-4-nitrobenzene ≥99.0% (by GC)
1-Iodo-4-nitrobenzene
Formule: C₆H₄INO₂
Kookpunt: 289 °C (1013 hPa)
Smeltpunt: 168 °C
Dichtheid: 2,273 g/cm³ (20 °C)
MDL Nummer: MFCD00007299
CAS nummer: 636-98-6
EINECS: 211-272-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e7a2d2592f6100c4ddecea46b1aca194639049d&vid=5e7a2d2592f6100c4ddecea46b1aca194639049d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 99.0 %
Melting point 172.0 to 176.0 deg-C
Solubility in hot Toluene almost transparency