1-Iodo-2,4-dinitrobenzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2,4-Dinitroiodobenzene, 1,3-Dinitro-4-iodobenzene

D1921-25G D1921-5G
TCIAD1921-25GEA 318 EUR
TCIAD1921-25G TCIAD1921-5G
1-Iodo-2,4-dinitrobenzene ≥98.0% (by GC)
1-Iodo-2,4-dinitrobenzene
Formule: C₆H₃IN₂O₄
MDL Nummer: MFCD00039738
CAS nummer: 709-49-9
EINECS: 211-908-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68f511977d79e99d6ab80ceefe3341be254b7da8&vid=96da883b65c1d11e1284d5b0b523d6660d4b1af3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR