4-Iodophenol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
I0311-10G I0311-25G
TCIAI0311-10GEA 40.7 EUR
TCIAI0311-10G TCIAI0311-25G
4-Iodophenol ≥98.0% (by GC)
4-Iodophenol
Formule: C₆H₅IO
MW: 220.01 g/mol
Kookpunt: 287
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002327
CAS nummer: 540-38-5
EINECS: 208-745-5
Merck Index: 14,05036
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9b6cef5729c2f4eeb6c1cdf6a2cbc457056c2c3&vid=9cd4b86ebb06a4ba3f1ceb4f3dd86f44415e1181&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 94.0 deg-C