N,N-Diethylaniline ≥99.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D0475-25ML D0475-500ML
TCIAD0475-25MLEA 22 EUR
TCIAD0475-25ML TCIAD0475-500ML
N,N-Diethylaniline ≥99.0% (by GC, titration analysis)
N,N-Diethylaniline
Formule: C₁₀H₁₅N
Kookpunt: 216°C
Dichtheid: 0,9348 g/cm³ (20 °C)
Vlampunt: 92°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009042
CAS nummer: 91-66-7
EINECS: 202-088-8
UN: 2432
ADR: 6.1,III
Merck Index: 14,03114
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b93b00d7f5488f6746219789d122490d0d267959&vid=e0d210652a8e75275d5b408285d82096391d9284&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR