4-Nitroanisool ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
N0126-25G N0126-500G N0126-100G
TCIAN0126-25GEA 17.6 EUR
TCIAN0126-25G TCIAN0126-500G TCIAN0126-100G
4-Nitroanisool ≥98.0% (by GC)
4-Nitroanisool
Formule: C₇H₇NO₃
Kookpunt: 274°C
Dichtheid: 1,254 g/cm³ (20 °C)
MDL Nummer: MFCD00007327
CAS nummer: 100-17-4
EINECS: 202-825-3
UN: 3458
ADR: 6.1,III
Merck Index: 14,06584
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b1aa84a00db3bf24653bf33a0c2ac555a94a3a8&vid=443d21b20f206a91b47f6dee9c47ce8bdcc2876a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown to Dark green powder to lump
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C
Solubility in Methanol almost transparency