2-Nitrofenol ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

N0219-25G N0219-500G N0219-100G
TCIAN0219-25GEA 17.6 EUR
TCIAN0219-25G TCIAN0219-500G TCIAN0219-100G
2-Nitrofenol ≥98.0% (by GC, titration analysis)
2-Nitrofenol
Formule: C₆H₅NO₃
Kookpunt: 216°C
Smeltpunt: 43…45 °C
Dichtheid: 1,599 g/cm³ (20 °C)
Vlampunt: 108 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011688
CAS nummer: 88-75-5
EINECS: 201-857-5
UN: 1663
ADR: 6.1,III
Merck Index: 14,06619
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9081cda9817d1cfd79fff2b533073e2a5ba2f694&vid=0d639cc33619de5d382bb8be56346a52258beb62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 44.0 to 47.5 deg-C
Solubility in EtOH (95vol%) almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR