4-Nitrofenol ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
N0220-25G
N0220-500G
N0220-100G
TCIAN0220-25GEA
20.9
EUR
TCIAN0220-25G
TCIAN0220-500G
TCIAN0220-100G
4-Nitrofenol ≥99.0% (by GC)
4-Nitrofenol
Formule:
C₆H₅NO₃ Kookpunt: 279°C Smeltpunt: 110…114 °C Dichtheid: 1,48 g/cm³ (20 °C) Vlampunt: 169 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007331 CAS nummer: 100-02-7 EINECS: 202-811-7 UN: 1663 ADR: 6.1,III Merck Index: 14,06620 |
|
Specification Test Results
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 112.0 to 116.0 deg-C |
Solubility in EtOH | almost transparency |
Transition interval (pH) | (very pale yellow)4.8 to 7.6 (yellow) |