2-Ethoxyphenol ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

E0230-25G E0230-500G
TCIAE0230-25GEA 17.6 EUR
TCIAE0230-25G TCIAE0230-500G
2-Ethoxyphenol ≥99.0% (by GC)
2-Ethoxyphenol
Formule: C₈H₁₀O₂
Kookpunt: 216°C
Smeltpunt: 28…29 °C
Dichtheid: 1,1 g/cm³ (20 °C)
Vlampunt: 86 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002187
CAS nummer: 94-71-3
EINECS: 202-358-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09c6e50dde350844c2c513e567de2b6ea010c2c3&vid=6b3a4113d0dd93d2d2e6c0ef0b06c968e8382629&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR