Ethyl-4-bromobutyrate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Ethyl 4-bromobutyrate
B0999-25G
B0999-500G
B0999-100G
TCIAB0999-25GEA
17.6
EUR
TCIAB0999-25G
TCIAB0999-500G
TCIAB0999-100G
Ethyl-4-bromobutyrate ≥97.0% (by GC)
Ethyl-4-bromobutyrate
Formule:
C₆H₁₁BrO₂ MW: 195.06 g/mol Kookpunt: 200°C Vlampunt: 58°C |
MDL Nummer:
MFCD00000259 CAS nummer: 2969-81-5 EINECS: 221-005-6 UN: 3272 ADR: 3 III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...