(S)-(+)-2-Aminobutyric acid ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: H-Abu-OH, L-alpha-Amino-N-Butyric acid, L-2-Aminobutyric acid, (S)-2-Aminobutanoic acid

A0826-1G A0826-5G
TCIAA0826-1GEA 23.1 EUR
TCIAA0826-1G TCIAA0826-5G
(S)-(+)-2-Aminobutyric acid ≥99.0% (by titrimetric analysis)
(S)-(+)-2-Aminobutyric acid
Formule: C₄H₉NO₂
MW: 103.12 g/mol
MDL Nummer: MFCD00064415
CAS nummer: 1492-24-6
EINECS: 216-083-3
Merck Index: 14,00428
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e28cb5798ee2357edc4607138873dde809ed57f&vid=0e28cb5798ee2357edc4607138873dde809ed57f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D +18.5 to +22.0 deg (C=5, HCl (2+1))
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR