Isoamyl butyrate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Isopentyl butyrate, Isoamyl Butyrate
B0760-25ML
B0760-500ML
TCIAB0760-25MLEA
17.6
EUR
TCIAB0760-25ML
TCIAB0760-500ML
Isoamyl butyrate ≥98.0% (by GC)
Isoamyl butyrate
Formule:
C₉H₁₈O₂ MW: 158.24 g/mol Vlampunt: 65°C |
MDL Nummer:
MFCD00044888 CAS nummer: 106-27-4 EINECS: 203-380-8 UN: 2620 ADR: 3 III Merck Index: 14,05115 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...