3-Chloroanilinium chloride ≥99.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3-Chloroaniline hydrochloride

C0113-25G C0113-500G
TCIAC0113-25GEA 17.6 EUR
TCIAC0113-25G TCIAC0113-500G
3-Chloroanilinium chloride ≥99.0% (by HPLC, total nitrogen)
3-Chloroanilinium chloride
Formule: C₆H₇Cl₂N
MW: 164.03 g/mol
MDL Nummer: MFCD00035485
CAS nummer: 141-85-5
EINECS: 205-506-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28b83d55303827d7d6c3b655dac01577a97cf472&vid=28b83d55303827d7d6c3b655dac01577a97cf472&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 99.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR