Diethyl bromomalonate ≥85.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B1205-25G B1205-500G
TCIAB1205-25GEA 24.2 EUR
TCIAB1205-25G TCIAB1205-500G
Diethyl bromomalonate ≥85.0% (by GC)
Diethyl bromomalonate
Formule: C₇H₁₁BrO₄
Kookpunt: 235°C
Vlampunt: 111°C
MDL Nummer: MFCD00009138
CAS nummer: 685-87-0
EINECS: 211-683-1
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c129fb44c341f7743070faaa057287b8969486af&vid=c129fb44c341f7743070faaa057287b8969486af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR