4-Amino-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
4-Cyano-3-trifluoromethylaniline
A1213-1G
A1213-5G
TCIAA1213-1GEA
20.9
EUR
TCIAA1213-1G
TCIAA1213-5G
4-Amino-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Amino-2-(trifluoromethyl)benzonitrile
Formule:
C₈H₅F₃N₂ MW: 186.14 g/mol |
MDL Nummer:
MFCD00042155 CAS nummer: 654-70-6 UN: 3439 ADR: 6.1 III |
|
Specification Test Results
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 141.0 to 144.0 deg-C |
Solubility in Pyridine | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...