Diphenyl sulphoxide ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Phenyl sulphoxide, sulphinyldibenzene
D1002-100G
D1002-25G
D1002-5G
TCIAD1002-100GEA
99
EUR
TCIAD1002-100G
TCIAD1002-25G
TCIAD1002-5G
Diphenyl sulphoxide ≥99.0% (by GC)
Diphenyl sulphoxide
Formule:
C₁₂H₁₀OS Kookpunt: 198°C |
MDL Nummer:
MFCD00002085 CAS nummer: 945-51-7 EINECS: 213-415-9 |
|
Specification Test Results
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 99.0 % |
Melting point | 69.0 to 72.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...