Ethyl isothiocyanatoformate ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

E0440-25G E0440-5G
TCIAE0440-25GEA 187 EUR
TCIAE0440-25G TCIAE0440-5G
Ethyl isothiocyanatoformate ≥95.0% (by GC)
Ethyl isothiocyanatoformate
Formule: C₄H₅NO₂S
Kookpunt: 61°C
Vlampunt: 50°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004814
CAS nummer: 16182-04-0
EINECS: 240-318-9
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60d2b5eb13b5791eb873bcbfec0dce5b6e6147e8&vid=60d2b5eb13b5791eb873bcbfec0dce5b6e6147e8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR