Thiomorpholine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
T1007-25ML
T1007-5ML
TCIAT1007-25MLEA
150
EUR
TCIAT1007-25ML
TCIAT1007-5ML
Thiomorpholine ≥98.0% (by GC)
Thiomorpholine
Formule:
C₄H₉NS Kookpunt: 169°C Vlampunt: 63°C |
MDL Nummer:
MFCD00005974 CAS nummer: 123-90-0 EINECS: 204-660-2 UN: 3267 ADR: 8,III Merck Index: 14,09300 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Specific gravity (20/20) | 1.0890 to 1.0930 |
Refractive index n20/D | 1.5370 to 1.5400 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...