Allylbenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Phenyl-2-propene, 3-Phenylpropene
P0609-25ML P0609-100ML
TCIAP0609-25MLEA 42.9 EUR
TCIAP0609-25ML TCIAP0609-100ML
Allylbenzene ≥98.0% (by GC)
Allylbenzene
Formule: (C₆H₅)CH₂CHCH₂
Kookpunt: 155°C
Smeltpunt: < –40 °C
Dichtheid: 0,89 g/cm³ (20 °C)
Vlampunt: 33°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008651
CAS nummer: 300-57-2
EINECS: 206-095-7
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd254d3885c7592cb5d34f22391a4efade39be74&vid=dd254d3885c7592cb5d34f22391a4efade39be74&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %