Phenethyl bromide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (2-Bromoethyl)benzene, 2-Phenylethyl bromide

B0594-25G B0594-500G B0594-100G
TCIAB0594-25GEA 18.7 EUR
TCIAB0594-25G TCIAB0594-500G TCIAB0594-100G
Phenethyl bromide ≥98.0% (by GC)
Phenethyl bromide
Formule: BrCH₂CH₂C₆H₅
Kookpunt: 220°C
Smeltpunt: –56 °C
Dichtheid: 1,37 g/cm³ (20 °C)
Vlampunt: 96°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000240
CAS nummer: 103-63-9
EINECS: 203-130-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f1e02460a0ad524b7db76d2c91b1000373c8668&vid=77f0dd7593c12fc92c78f84aa49717d00d1600a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR