4-tert-Butyl-1-bromobenzene ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 1-Bromo-4-tert-butylbenzene, 2-(4-Bromophenyl)-2-methylpropane, 1-Bromo-4-(tert-butyl)benzene, 4-Bromo-tert-butylbenzene
B1727-100G B1727-25G
TCIAB1727-100GEA 53.9 EUR
TCIAB1727-100G TCIAB1727-25G
4-tert-Butyl-1-bromobenzene ≥96.0% (by GC)
4-tert-Butyl-1-bromobenzene
Formule: C₁₀H₁₃Br
Kookpunt: 228°C
Dichtheid: 1,2572 g/cm³ (15 °C)
Vlampunt: 97°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000108
CAS nummer: 3972-65-4
EINECS: 223-599-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29fc420c81fcd7fd3a50ac4ed47d4305ff3c6855&vid=30a0c2844aba419d371b85ab3e0e8334ac9fb6ba&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 96.0 %