4-Bromotoluene ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B0661-25G B0661-500G B0661-100G
TCIAB0661-25GEA 30.8 EUR
TCIAB0661-25G TCIAB0661-500G TCIAB0661-100G
4-Bromotoluene ≥99.0% (by GC)
4-Bromotoluene
Formule: C₇H₇Br
Kookpunt: 184°C
Smeltpunt: 24…27 °C
Dichtheid: 1,387 g/cm³ (20 °C)
Vlampunt: 85 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000109
CAS nummer: 106-38-7
EINECS: 203-391-8
UN: 3077
ADR: 9,III
Merck Index: 13,01424
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bfff899c2ae74501b4ba8a41c556ba48f882259&vid=0382cf87239f26e08163d63226685b14caceb70f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 99.0 %
Solubility in Methanol almost transparency