2-Bromo-6-chlorotoluene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-Bromo-3-chloro-2-methylbenzene
B2032-25G B2032-5G
TCIAB2032-25GEA 79.2 EUR
TCIAB2032-25G TCIAB2032-5G
2-Bromo-6-chlorotoluene ≥98.0% (by GC)
2-Bromo-6-chlorotoluene
Formule: C₇H₆BrCl
Kookpunt: 120°C
Vlampunt: 95°C
MDL Nummer: MFCD00061121
CAS nummer: 62356-27-8
EINECS: 263-520-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b10cb249ad3bd87d21aafab4eefbdf2c5e08bb1b&vid=7f65d7f11f9ef5692b5a9eb2a38c255d081fd255&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light red to Green clear liquid
Purity (GC) min. 98.0 %