5-Bromo-2-chlorobenzotrifluoride ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C1846-25G C1846-5G
TCIAC1846-25GEA 58.3 EUR
TCIAC1846-25G TCIAC1846-5G
5-Bromo-2-chlorobenzotrifluoride ≥98.0% (by GC)
5-Bromo-2-chlorobenzotrifluoride
Formule: C₇H₃BrClF₃
Kookpunt: 195°C
Smeltpunt: -21°C
Dichtheid: 1,7486 g/cm³ (25 °C)
Vlampunt: 81°C
MDL Nummer: MFCD00000601
CAS nummer: 445-01-2
EINECS: 207-149-2

Order Now

Specification Test Results

Appearance White to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR