α-Bromo-3-nitrotoluene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

N0625-10G N0625-25G
TCIAN0625-10GEA 17.6 EUR
TCIAN0625-10G TCIAN0625-25G
α-Bromo-3-nitrotoluene ≥98.0% (by GC)
α-Bromo-3-nitrotoluene
Formule: C₇H₆BrNO₂
Kookpunt: 115°C
Smeltpunt: 59…60 °C
MDL Nummer: MFCD00007271
CAS nummer: 3958-57-4
EINECS: 223-557-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c70587e5f0602af2b1226a6870432fdca12993a1&vid=c78b14295b13d147b63adf8910d7ead7cfcf8790&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR