2-Bromo-4-nitrotoluene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B1262-25G B1262-250G
TCIAB1262-25GEA 34.1 EUR
TCIAB1262-25G TCIAB1262-250G
2-Bromo-4-nitrotoluene ≥98.0% (by GC)
2-Bromo-4-nitrotoluene
Formule: C₇H₆BrNO₂
Kookpunt: 175°C
MDL Nummer: MFCD00007195
CAS nummer: 7745-93-9
EINECS: 231-809-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a625e98219f0ab4f3864d8410100a8d6de05d538&vid=93d88f92d28895c78a186ee104cc39043b0969bd&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C
Solubility in hot Methanol very faint turbidity