2-Tolualdehyde ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T0566-25G T0566-500G T0566-100G
TCIAT0566-25GEA 37.4 EUR
TCIAT0566-25G TCIAT0566-500G TCIAT0566-100G
2-Tolualdehyde ≥98.0% (by GC)
2-Tolualdehyde
Formule: C₈H₈O
Kookpunt: 200°C
Smeltpunt: –35 °C
Dichtheid: 1,0324 g/cm³ (20 °C)
Vlampunt: 67°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003338
CAS nummer: 529-20-4
EINECS: 208-452-2
Merck Index: 14,09527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce8e4a9e7605c01ea16f1cf58a9735b1a02d36be&vid=d926b7449fda11f0ace6349d6cccc6440f8b1fdc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR