4-Butylbenzaldehyde ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B1014-25ML
B1014-5ML
TCIAB1014-25MLEA
323
EUR
TCIAB1014-25ML
TCIAB1014-5ML
4-Butylbenzaldehyde ≥95.0% (by GC)
4-Butylbenzaldehyde
Formule:
C₁₁H₁₄O MW: 162.23 g/mol Kookpunt: 165°C Smeltpunt: 43 Vlampunt: 128°C |
MDL Nummer:
MFCD00040751 CAS nummer: 1200-14-2 EINECS: 214-851-2 |
|
Specification Test Results
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...