4-Tolualdehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
p-Tolualdehyde, p-Tolylaldehyde, p-Methylbenzaldehyd, 4-Methylbenzaldehyd, 4-Methylbenzaldehyde, 4-Tolylaldehyde
T0259-25ML
T0259-500ML
T0259-100ML
TCIAT0259-25MLEA
27.5
EUR
TCIAT0259-25ML
TCIAT0259-500ML
TCIAT0259-100ML
4-Tolualdehyde ≥98.0% (by GC)
4-Tolualdehyde
Formule:
C₈H₈O MW: 120.15 g/mol Kookpunt: 204°C Smeltpunt: -6 Vlampunt: 88°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006954 CAS nummer: 104-87-0 EINECS: 203-246-9 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...